2,2'-Bipyridyl ACS, GR for analysis, reagent for iron(II) and molybdenum, Supelco®

Supplier: Merck
Danger

Synonyms: α,α′-Dipyridyl, 2,2'-Bipyridine, 2,2'-Dipyridyl

M-Clarity™ quality level = MQ100

1.03098.0025
1.03098.0025EA 277 EUR
1.03098.0025
2,2'-Bipyridyl ACS, GR for analysis, reagent for iron(II) and molybdenum, Supelco®
2,2'-Bipyridyl
Formula: C₁₀H₈N₂
MW: 156.19 g/mol
Boiling Pt: 273
Melting Pt: 70…72 °C
Density: 1.326 g/cm³ (–150 °C)
Flash Pt: 121
Storage Temperature: Ambient
MDL Number: MFCD00006212
CAS Number: 366-18-7
EINECS: 206-674-4
UN: 2811
ADR: 6.1 III
Merck Index: 13,03382
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e86bbc29edb6c5d09c058b8efcb6b9eb7f4d070c&vid=9e3a03e37045643109ae0765802e1997d30604ae&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (perchloric acid titration) ≥ 99.5 %
Identity (IR-spectrum) passes test
Melting point 70 - 72 °C
Absorbance of solution (50% w/v; methanol) conforms
Sensitivity (to ferrous ion) passes test
Flash Point 121 °C
pH 7.5 (5 g/l, H2O, 25 °C)
Hazard Class 6.1
Hazard Class Description Toxic materials


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR