Dimethyl sulphoxide-[D₆] ≥99.8% (Isotopic), MagniSolv™ for NMR spectroscopy, Sigma-Aldrich®

Supplier: Merck

Synonyms: Dimethyl sulphoxide-[²H₆], Hexadeuterodimethyl sulphoxide, DMSO-D6, DMSO-d6, Dimethyl sulphoxide-d6

M-Clarity™ quality level = MQ100

1.03424.0009 1.03424.0010 1.03424.0100 1.03424.0025 1.03424.0005 1.03424.0011 1.03424.0050
1.03424.0009EA 61.8 EUR
1.03424.0009 1.03424.0010 1.03424.0100 1.03424.0025 1.03424.0005 1.03424.0011 1.03424.0050
Dimethyl sulphoxide-[D₆] ≥99.8% (Isotopic), MagniSolv™ for NMR spectroscopy, Sigma-Aldrich®
Dimethyl sulphoxide-[D6]
Formula: D₃CS(O)CD₃
MW: 84.09 g/mol
Boiling Pt: 189
Melting Pt: 20.2
Density: 1.19 g/cm³ (20 °C)
Flash Pt: 82 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00002090
CAS Number: 2206-27-1
EINECS: 218-617-0

Order Now

Specification Test Results

Deuteration degree ≥ 99.80 %
Water (K. Fischer, H₂O + D₂O) ≤ 0.030 %
Performance test (NMR-spectrum) passes test
Water (NMR, H₂O) ≤ 0.020 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR