Sodium diphenylamine-4-sulphonate, Millipore®

Supplier: Merck

Synonyms: Sodium Diphenylaminesulphonate, Sodium N-phenylsulphanilate, Sodium (4-anilinobenzenesulphonate), Diphenylamine-4-sulphonic acid, sodium salt

M-Clarity™ quality level = MQ200

1.03590.0010
1.03590.0010EA 308 EUR
1.03590.0010
Sodium diphenylamine-4-sulphonate, Millipore®
Sodium diphenylamine-4-sulphonate
Formula: C₁₂H₁₀NNaO₃S
Storage Temperature: Ambient
MDL Number: MFCD00036468
CAS Number: 6152-67-6
EINECS: 228-165-6
Merck Index: 13,07397

Order Now

Specification Test Results

Solubility 820 g/l (20 °C)
Melting Point 200 °C
Molar Mass 271.26 g/mol
Bulk Density 160 kg/m3
pH 5.7 (10 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR