Dimethyl sulphoxide-[D₆] (99.8% D) + 0.03% v/v TMS, MagniSolv™ for NMR spectroscopy, Sigma-Aldrich®

Supplier: Merck

Synonyms: Dimethyl sulphoxide-[²H₆], Hexadeuterodimethyl sulphoxide, DMSO-D6, DMSO-d6, Dimethyl sulphoxide-d6

M-Clarity™ quality level = MQ100

1.03591.0100 1.03591.0050
1.03591.0100EA 478 EUR
1.03591.0100 1.03591.0050
Dimethyl sulphoxide-[D₆] (99.8% D) + 0.03% v/v TMS, MagniSolv™ for NMR spectroscopy, Sigma-Aldrich®
Dimethyl sulphoxide-[D6]
Formula: D₃CS(O)CD₃
MW: 84.09 g/mol
Boiling Pt: 189
Melting Pt: 20.2
Density: 1.19 g/cm³ (20 °C)
Flash Pt: 82 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00002090
CAS Number: 2206-27-1
EINECS: 218-617-0

Order Now

Specification Test Results

Deuteration degree ≥ 99.8 %
Water (K. Fischer, H₂O + D₂O) ≤ 0.030 %
Performance test (NMR-spectrum) passes test
Water (NMR, H₂O) ≤ 0.020 %
Tetramethylsilane (NMR-spectrum) 0.03%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR