HPTLC plates with spherical particles, LiChrospher®
Supplier: Merck
1.05586.0001 1.05646.0001 1.15445.0001
1.05586.0001EA 500 EUR
1.05586.0001 1.05646.0001 1.15445.0001
HPTLC plates with spherical particles, LiChrospher®
TLC Plates
HPTLC LiChrospher® plates are based on spherical-shaped silica 60 with a particle size of 7 µm, and narrow particle size distribution similar to the phases used in HPLC. LiChrospher® HPTLC plates possess a selectivity that is comparable to respective HPTLC plates, however, plate height and separation numbers are further improved resulting in shorter analysis times.
  • 20% reduced running times
  • Highly compact spots or zones
  • Lower detection limits and better resolution
LiChrospher® HPTLC plates are especially suitable for separations of highly complex, lowMore Product Information