Sodium hydroxide 10% in aqueous solution, EMSURE® for analysis, Supelco®

Supplier: Merck
Danger


M-Clarity™ quality level = MQ300

1.05588.1000 1.05588.9010
1.05588.1000EA 36.29 EUR
1.05588.1000 1.05588.9010
Sodium hydroxide 10% in aqueous solution, EMSURE® for analysis, Supelco®
Sodium hydroxide
Formula: NaOH
Boiling Pt: 119…135 °C (1013 hPa)
Melting Pt: 9…10 °C
Density: 1,53 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8,II

Order Now

Specification Test Results

Solubility (20 °C) soluble
Density 1.11 g/cm3 (20 °C)
pH 13.8 (H2O, 20 °C)
Hazard Class 8
Hazard Class Description Corrosive Material


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR