Lithium carbonate, Suprapur® for trace analysis, Supelco®

Supplier: Merck
Warning


M-Clarity™ quality level = MQ100

1.05676.0250
1.05676.0250EA 2010 EUR
1.05676.0250
Lithium carbonate, Suprapur® for trace analysis, Supelco®
Lithium carbonate
Formula: Li₂CO₃
Melting Pt: 720 °C
Density: 2,11 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011084
CAS Number: 554-13-2
EINECS: 209-062-5
Merck Index: 13,05549

Order Now

Specification Test Results

Solubility 13 g/l (20 °C)
Melting Point 720 °C
Molar Mass 73.89 g/mol
Bulk Density 250 kg/m3
Vapor Pressure (20 °C)
Density 2.1 g/cm3 (20 °C)
pH 10 - 11 (5 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR