Lithium sulphate monohydrate, EMSURE® Reag. Ph. Eur., ACS analysis_for, Supelco®

Supplier: Merck
Warning


MQ300

1.05694.0250
1.05694.0250EA 208 EUR
1.05694.0250
Lithium sulphate monohydrate, EMSURE® Reag. Ph. Eur., ACS analysis_for, Supelco®
Lithium sulphate monohydrate
Formula: Li₂SO₄·H₂O
MW: 127.96 g/mol
Melting Pt: 120
Density: 2.05 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149766
CAS Number: 10102-25-7
EINECS: 233-820-4

Order Now

Specification Test Results

Solubility 340 g/l (20 °C)
Melting Point 120 °C
Molar Mass 127.96 g/mol
Bulk Density 940 kg/m3
Density 2.05 g/cm3 (20 °C)
pH 4.5 - 6.0 (50 g/l, H2O, 20 °C)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR