with moisture indicator ~10 mesh, Ø ~2 mm, Supelco®

Supplier: Merck

Synonyms: Molecular sieve 0.3 nm, 3A molecular sieve, Molecular sieve A3, Linde 3A, Sodium aluminium silicate

MQ200

1.05734.0250 1.05734.1000
1.05734.0250EA 154 EUR
1.05734.0250 1.05734.1000
with moisture indicator ~10 mesh, Ø ~2 mm, Supelco®
Molecular sieve 3A (0.3 nm, 3 Å)
Formula: KnNa₁₂-n[(AlO₂)₁₂(SiO₂)₁₂]·xH₂O
MDL Number: MFCD00131613
CAS Number: 308080-99-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR