L(-)-Menthol, crystallised, EMPROVE® ESSENTIAL Ph. Eur., BP, JP, USP, SAFC®

Supplier: MERCK PRODUCTION CHEMICALS
Warning

1.05995.1000 1.05995.9025
1.05995.1000EA 497 EUR
1.05995.1000 1.05995.9025
L(-)-Menthol, crystallised, EMPROVE® ESSENTIAL Ph. Eur., BP, JP, USP, SAFC®
L(-)-Menthol
Formula: C₁₀H₂₀O
Boiling Pt: 212 °C (1013 hPa)
Melting Pt: 41…44 °C
Density: 0,89 g/cm³ (20 °C)
Flash Pt: 96 °C
Storage Temperature: Ambient
MDL Number: MFCD00062979
CAS Number: 2216-51-5
EINECS: 218-690-9
Merck Index: 13,05861

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR