β-(N-Morpholino)ethanesulphonic acid (MES) monohydrate, Millipore®

Supplier: Merck


M-Clarity™ quality level = MQ100

1.06126.0025 1.06126.0250 1.06126.9010 1.06126.1000
1.06126.0025EA 52.7 EUR
1.06126.0025 1.06126.0250 1.06126.9010 1.06126.1000
β-(N-Morpholino)ethanesulphonic acid (MES) monohydrate, Millipore®
β-(N-Morpholino)ethanesulphonic acid (MES) monohydrate
Formula: C₆H₁₃NO₄S·1H₂O
Melting Pt: > 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00149409
CAS Number: 145224-94-8
EINECS: 224-632-3
Merck Index: 13,05929

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR