4-Methyl-2-pentanone, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®

Supplier: Merck
Danger

Synonyms: Methyl iso-Butyl Ketone, Isobutyl Methyl Ketone, Methyl Isobutyl Ketone, MIK

M-Clarity™ quality level = MQ300

1.06146.1000 1.06146.2500 1.06146.6025
1.06146.1000EA 57.15 EUR
1.06146.1000 1.06146.2500 1.06146.6025
4-Methyl-2-pentanone, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
4-Methyl-2-pentanone
Formula: (CH₃)₂CHCH₂COCH₃
MW: 100.16 g/mol
Boiling Pt: 117.4
Melting Pt: –84 °C
Density: 0.801 g/cm³ (20 °C)
Flash Pt: 16
Storage Temperature: Ambient
MDL Number: MFCD00008938
CAS Number: 108-10-1
EINECS: 203-550-1
UN: 1245
ADR: 3 II
Merck Index: 13,05227
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3d44947ef43cbc8103ae523ce5da380ef184ba09&vid=373b9075524004b95d32d3cf9698896714467e8c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 18 - 20 g/l (20 °C)
Melting Point -84 °C
Molar Mass 100.16 g/mol
Boiling Point 116 - 118 °C (1013 hPa)
Vapor Pressure 20 hPa (20 °C)
Flash Point 14 °C
Refractive Index 1.3962 (20 °C)
Explosion Limit 1.2 - 8.0 %(V)
Density 0.80 g/cm3 (20 °C)
pH (H2O, 20 °C) neutral
Ignition Point 460 °C
Evaporation number 10


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR