Sodium bromide, EMPROVE® ESSENTIAL Ph. Eur., BP, JP, USP, SAFC®

Supplier: MERCK PRODUCTION CHEMICALS

1.06360.9025
1.06360.9025EA 0 EUR
1.06360.9025
Sodium bromide, EMPROVE® ESSENTIAL Ph. Eur., BP, JP, USP, SAFC®
Sodium bromide
Formula: NaBr
Boiling Pt: 1396 °C (1013 hPa)
Melting Pt: 753 °C
Density: 3,2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003475
CAS Number: 7647-15-6
EINECS: 231-599-9
Merck Index: 13,08666

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR