Sodium borohydride analytical reagent, Supelco®

Supplier: Merck
Danger

Synonyms: Sodium tetrahydridoborate, Sodium tetrahydroborate

M-Clarity™ quality level = MQ100

1.06371.0100
1.06371.0100EA 359 EUR
1.06371.0100
Sodium borohydride analytical reagent, Supelco®
Sodium borohydride
Formula: NaBH₄
MW: 37.83 g/mol
Melting Pt: 400
Density: 1.07 g/cm³ (20 °C)
Flash Pt: 69
Storage Temperature: Ambient
MDL Number: MFCD00003518
CAS Number: 16940-66-2
EINECS: 241-004-4
UN: 1426
ADR: 4.3 I
Merck Index: 13,08664

Order Now

Specification Test Results

Solubility 550 g/l (25 °C) (slow decomposition)
Melting Point 400 °C (slow decomposition)
Molar Mass 37.83 g/mol
Bulk Density 350 - 500 kg/m3
Boiling Point 500 °C (decomposition)
Explosion Limit 3.02 %(V)
Density 1.07 g/cm3 (20 °C)
pH 11 (10 g/l, H2O, 20 °C)
Ignition Point 220 °C
Hazard Class 4.3
Hazard Class Description Dangerous When Wet Material


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR