Sodium chlorate, Supelco®

Supplier: Merck
Danger


M-Clarity™ quality level = MQ200

1.06420.1000 1.06420.9050
1.06420.1000EA 57.1 EUR
1.06420.1000 1.06420.9050
Sodium chlorate, Supelco®
Sodium chlorate
Formula: ClNaO₃
MW: 106.44 g/mol
Boiling Pt: 300
Melting Pt: 260
Density: 2.498 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003479
CAS Number: 7775-09-9
EINECS: 231-887-4
UN: 1495
ADR: 5.1 II
REACH: 01-2119474389-23
Merck Index: 13,08670

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR