Standard reference materials and buffer solutions, CertiPUR®, Supelco®
Supplier: Merck
1.01960.0001 1.01961.0025 1.01963.0025 1.01964.0025 1.01965.0025 1.07200.0105 1.07202.0105 1.07204.0105 1.07205.0105 1.07203.0105 1.01962.0001
1.01960.0001EA 421 EUR
1.01960.0001 1.01961.0025 1.01963.0025 1.01964.0025 1.01965.0025 1.07200.0105 1.07202.0105 1.07204.0105 1.07205.0105 1.07203.0105 1.01962.0001
Standard reference materials and buffer solutions, CertiPUR®, Supelco®
Buffers pH Calibration Buffers
Materials for the precise calibration and monitoring of pH instruments.
  • Traceable to NIST primary standard and PTB
  • Maximum deviation ±0,003 pH units
  • Measured in an independent accredited calibration laboratory for the pH value measurement (DKD-K-14301)