Lead(II) nitrate, EMSURE® Reag. Ph. Eur., ACS analytical reagent, Supelco®

Supplier: Merck
Danger

Synonyms: Lead dinitrate

M-Clarity™ quality level = MQ300

1.07398.0100 1.07398.1000
1.07398.0100EA 63.1 EUR
1.07398.0100 1.07398.1000
Lead(II) nitrate, EMSURE® Reag. Ph. Eur., ACS analytical reagent, Supelco®
Lead (II) nitrate
Formula: Pb(NO₃)₂
MW: 331.21 g/mol
Melting Pt: 450…470 °C
Density: 4.53 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011153
CAS Number: 10099-74-8
EINECS: 233-245-9
UN: 1469
ADR: 5.1 (6.1) II
Merck Index: 13,05432

Order Now

Specification Test Results

Solubility 522 g/l (20 °C)
Melting Point 458 - 459 °C
Molar Mass 331.2 g/mol
Bulk Density 1850 kg/m3
Vapor Pressure (20 °C)
Density 4.49 g/cm3 (20 °C)
pH 3 - 4 (50 g/l, H2O, 20 °C)
Hazard Class 5.1
Hazard Class Description Oxidizer

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR