Lead(II) acetate basic, anhydrous, EMSURE® Sugar_analysis Acc. To Horne, Supelco®

Supplier: Merck
Danger

Synonyms: Horne′s compound, Lead(II) hydroxide acetate, Acetic acid lead (II) salt basic, Lead subacetate

MQ300

1.07414.1000 1.07414.9030
1.07414.1000EA 91.7 EUR
1.07414.1000 1.07414.9030
Lead(II) acetate basic, anhydrous, EMSURE® Sugar_analysis Acc. To Horne, Supelco®
Lead (II) acetate basic
Formula: (CH₃COO)₂Pb·2Pb(OH)₂
MW: 807.72 g/mol
Melting Pt: 198…205 °C
Density: 3.58 g/cm3 (22 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011154
CAS Number: 51404-69-4
EINECS: 215-630-3
UN: 2291
ADR: 6.1 III

Order Now

Specification Test Results

Assay (PbO) ≥ 33.0 %
In water insoluble matter ≤ 1.0 %
Substances insoluble in dilute acetic acid ≤ 0.02 %
Chloride (Cl) ≤ 0.003 %
Nitrate und Nitrite (as NO₃) passes test
Ca (Calcium) ≤ 0.01 %
Cu (Copper) ≤ 0.002 %
Fe (Iron) ≤ 0.002 %
K (Potassium) ≤ 0.02 %
Na (Sodium) ≤ 0.05 %
Loss on Drying (105 °C) ≤ 1.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR