Strontium nitrate, EMSURE® for analysis, Supelco®

Supplier: Merck
Danger


M-Clarity™ quality level = MQ300

1.07872.0250 1.07872.9025
1.07872.0250EA 116 EUR
1.07872.0250 1.07872.9025
Strontium nitrate, EMSURE® for analysis, Supelco®
Strontium nitrate
Formula: Sr(NO₃)₂
MW: 211.63 g/mol
Boiling Pt: 645
Melting Pt: 570
Density: 2.113 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011248
CAS Number: 10042-76-9
EINECS: 233-131-9
UN: 1507
ADR: 5.1 III
Merck Index: 13,08926

Order Now

Specification Test Results

Solubility 660 g/l (20 °C)
Melting Point 570 °C
Molar Mass 211.63 g/mol
Bulk Density 1000 kg/m3
Boiling Point 645 °C
Density 2.986 g/cm3 (20 °C)
pH 5 - 7 (50 g/l, H2O, 20 °C)
Hazard Class 5.1
Hazard Class Description Oxidizer


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR