Zinc, granules, EMSURE® ISO analytical reagent, Supelco®

Supplier: Merck
Warning


M-Clarity™ quality level = MQ300

1.08780.0500 1.08780.1000
1.08780.0500EA 161 EUR
1.08780.0500 1.08780.1000
Zinc, granules, EMSURE® ISO analytical reagent, Supelco®
Zinc

*1 AOX Enrichment Set  1.00678.0001 and AOX Sample Preparation Set 1.00677 required


*3 BOD Nutrient Salt Mixture  1.00687 required


*4 Reagents for free chlorine testing are Reagent Cl2-1 1.00086.0001  and Reagent Cl2-2 1.00087.0001 together with empty cell and accessories  1.00089.0001


Reagents for total chlorine are Reagent Cl2-1 1.00086.0001, Reagent Cl2-2 1.00087.0001 and Reagent Cl2-3 1.00088.0001  together with empty cell and accessories  1.00089.0001


*5 TOC screw caps for digestion required


*6 Zinc Reagent-6 required

Formula: Zn
Boiling Pt: 908 °C (1013 hPa)
Melting Pt: 420 °C
Density: 7,14 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011291
CAS Number: 7440-66-6
EINECS: 231-175-3
UN: 3077
ADR: 9,III
Merck Index: 12,10255

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point 419 °C
Molar Mass 65.39 g/mol
Boiling Point 906 °C (1013 hPa)
Density 7.13 g/cm3 (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR