Perchloric acid 0.1 mol/l (0.1 N) anhydrous acetic acid, Titripur® Reag. USP, Reag. Ph. Eur. Titration_nonaqueous_media, Supelco®

Supplier: Merck
Danger


MQ100

1.09065.1000 1.09065.1003
1.09065.1000EA 49.79 EUR
1.09065.1000 1.09065.1003
Perchloric acid 0.1 mol/l (0.1 N) anhydrous acetic acid, Titripur® Reag. USP, Reag. Ph. Eur. Titration_nonaqueous_media, Supelco®
Perchloric acid

Merck have introduced into the range of Titripur® Titrants some packs with additional technology - Supelco® SmartTitrants that use an RFID tag to provide a seamless data transfer between the smart titrator and the RFID tag on the label of the bottle of titrant. It will ensure that all information needed for method execution, result calculation and documentation is transferred to the titrator equipped with this technology. MettlerToledo Compact and Excellence titrators include this technology. Please see ordering table below for packs with RFID tag.

Formula: HClO₄
MW: 100.46 g/mol
Boiling Pt: 39
Melting Pt: -101
Density: 1.06 g/cm³ (20 °C)
Flash Pt: 40 °C
Storage Temperature: Ambient
MDL Number: MFCD00011325
CAS Number: 7601-90-3
EINECS: 200-580-7
UN: 2789
ADR: 8 (3) II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5fc15d8358229daccc96a56573b32589d2a21575&vid=3780c3029836d8b95181cdef71dc2669f7e2a797&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Form liquid
Amount-of-substance concentration 0.0995 - 0.1005
Measurement uncertainty +/- 0.0003
Water content (according to Karl Fischer) 0.1 - 0.2 %
Traceability NIST SRM
The concentration is determined by volumetric titration and refers to 20°C.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR