pH indicator strips, MQuant®, Supelco®
Supplier: Merck
1.09531.0001 1.09532.0001 1.09533.0001 1.09535.0001 1.09540.0001 1.09541.0001 1.09542.0001 1.09543.0001 1.09545.0001 1.09584.0001 1.09450.0010 1.09502.0001 1.09531.0003-P
1.09531.0001EA 17.53 EUR
1.09531.0001 1.09532.0001 1.09533.0001 1.09535.0001 1.09540.0001 1.09541.0001 1.09542.0001 1.09543.0001 1.09545.0001 1.09584.0001 1.09450.0010 1.09502.0001 MERC1.09531.0003-P
pH indicator strips, MQuant®, Supelco®
Test Papers pH Indicator Papers and Test Strips
These test strips contain special indicator dyes that are covalently bound to the reagent papers which are then adhered to the plastic strips.
  • Non bleeding - do not contaminate the solution to be measured
  • Quality assurance - tests are checked and calibrated against certified buffer solutions
  • Wide and narrow pH ranges available
Accessories information: Analyse your pH 0 to 14 test strips (1.09535) with this reference card and the free MQuant® StripScan app. The brilliant colour-fast reference colours allow for robust measurement results even underMore Product Information