Ethyl acetate, EMSURE® ACS, ISO, Reag. Ph. Eur. 1035300 for analysis, Supelco®

Supplier: Merck
Danger


M-Clarity™ quality level = MQ300

1.09623.1000 1.09623.2500 1.09623.6010 1.09623.6025 1.09623.9026 1.09623.9181 1.09623.4000 1.09623.5011
1.09623.1000EA 36.41 EUR
1.09623.1000 1.09623.2500 1.09623.6010 1.09623.6025 1.09623.9026 1.09623.9181 1.09623.4000 1.09623.5011
Ethyl acetate, EMSURE® ACS, ISO, Reag. Ph. Eur. 1035300 for analysis, Supelco®
Ethyl acetate

EMSURE® grade solvents are suitable for a broad spectrum of classical lab applications, and are frequently used in regulated and highly demanding lab applications. EMSURE® provides worldwide best and most extensive product specifications. We declare our EMSURE® range to be in compliance with the ACS, with the reagent part of the European Pharmacopoeia (Reag. Ph Eur) and also with the ISO standards.

Formula: CH₃COOC₂H₅
Boiling Pt: 77,1 °C (1013 hPa)
Melting Pt: –83 °C
Density: 0,902 g/cm³ (20 °C)
Flash Pt: –4 °C
Storage Temperature: Ambient
MDL Number: MFCD00009171
CAS Number: 141-78-6
EINECS: 205-500-4
UN: 1173
ADR: 3,
REACH: 01-2119475103-46
Merck Index: 13,03792
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=226aecffe42ffab125597552eff0ef7f88ef9983&vid=f5707ce0faee09ca10700419252c5841e0fbc01a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 85.3 g/l (20°C)
Melting Point -83 °C
Boiling Point 77 °C (1013 hPa)
Vapor Pressure 97 hPa (20 °C)
Flash Point -4 °C
Refractive Index 1.37
Explosion Limit 2.1 - 11.5 %(V)
Density 0.90 g/cm3 (20 °C)
Ignition Point 460°C
Grade ACS,ISO,Reag. Ph Eur
Synonyms Ethyl ethanoate, Acetic ester, Ethyl ester
Hill Formula C4H8O2
Chemical Formula CH3COOC2H5


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR