Sodium thiosulphate 0.01 mol/l concentrated aqueous solution, Titrisol®, Supelco®

Supplier: Merck

Synonyms: Sodiumthiosulphate, Sodium hyposulphite, Antichlor, di-Sodium thiosulphate

M-Clarity™ quality level = MQ100

1.09909.0001
1.09909.0001EA 20.33 EUR
1.09909.0001
Sodium thiosulphate 0.01 mol/l concentrated aqueous solution, Titrisol®, Supelco®
Sodium thiosulphate
Formula: Na₂S₂O₃
Storage Temperature: Ambient
MDL Number: MFCD00003499
CAS Number: 7772-98-7
EINECS: 231-867-5

Order Now

Specification Test Results

Solubility (20 °C) soluble
Density 1.02 g/cm3 (20 °C)
pH 9 - 10 (H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR