Zinc standard solution, concentrate, 1000 mg Zn in 0,06% hydrochloric acid (from ZnCl₂) Titrisol® standard for AAS, Supelco®
Supplier: Merck
1.09953.0001
1.09953.0001EA 37.8 EUR
1.09953.0001
Zinc standard solution, concentrate, 1000 mg Zn in 0,06% hydrochloric acid (from ZnCl₂) Titrisol® standard for AAS, Supelco®
Standards Cationic Standards Zinc Standards
Certifications: Traceable to SRM from NIST
Order Now
SPECIFICATIONS
Environmentally Preferable
M-Clarity quality level MQ100