Buffer tablets according to Weise, Sigma-Aldrich®
Supplier: Merck
1.09468.0100 1.11374.0100 1.11373.0100
1.09468.0100EA 111 EUR
1.09468.0100 1.11374.0100 1.11373.0100
Buffer tablets according to Weise, Sigma-Aldrich®
Buffers Microscopy Buffers
Ideal for preparing buffer solution according to WEISE for staining of blood smears.
Prepared buffer solutions are used in hematology, for the dilution of staining solutions and for the rinsing steps in the staining process. Each tablet is sufficient for one litre of buffer solution, each produced solution can be stable for up to 4 weeks.
Order Now
SPECIFICATIONS
Environmentally Preferable
M-Clarity quality level MQ300