Sudan III, Millipore®

Supplier: Merck

Synonyms: Solvent Red 23, Sudan® III (Red), Scarlet B, Sudan Red III, Cerasin Red, Sudan Red BK, Sudan Red B, Fat Ponceau G, Sudan G, D and C Red No. 17, Oil red, Tony Red, Oil scarlet, Organol® red BS, 1-(4-(Phenylazo)phenylazo)-2-naphthol, Sudan V, 1-((4-(Phenyldiazenyl)phenyl)diazenyl)naphthalen-2-ol

M-Clarity™ quality level = MQ200

1.11747.0025
1.11747.0025EA 98 EUR
1.11747.0025
Sudan III, Millipore®
Sudan III
Formula: C₂₂H₁₆N₄O
MW: 352.4 g/mol
Melting Pt: 205.3…206.3 °C
MDL Number: MFCD00003905
CAS Number: 85-86-9
EINECS: 201-638-4
Merck Index: 12,09054

Order Now

Specification Test Results

Melting Point 199 °C (decomposition)
Molar Mass 352.39 g/mol


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR