TLC and GLP plates, silica gel 60, unmodified layers, glass backed
Supplier: Merck
1.05626.0001 1.05714.0001 1.05715.0001 1.05719.0001 1.05721.0001 1.05724.0001 1.05729.0001 1.05789.0001 1.05808.0001 1.11798.0001 1.11844.0001 1.11845.0001 1.11846.0001 1.15326.0001 1.15341.0001 1.16485.0001 1.15327.0001
1.05626.0001EA 339 EUR
1.05626.0001 1.05714.0001 1.05715.0001 1.05719.0001 1.05721.0001 1.05724.0001 1.05729.0001 1.05789.0001 1.05808.0001 1.11798.0001 1.11844.0001 1.11845.0001 1.11846.0001 1.15326.0001 1.15341.0001 1.16485.0001 1.15327.0001
TLC and GLP plates, silica gel 60, unmodified layers, glass backed
TLC Plates
TLC plates, unmodified silica gel 60
Merck silica gel 60 has a unique polymeric binder that results in a uniform, hard surface that will not easily crack or blister. This smooth, extremely dense plate coating ensures narrow bands and maximum separation efficiency with the lowest background noise. Layer thickness 250 μm and particle size of 10 to 12 µm. Two inorganic fluorescent indicators for UV detection of colourless substances.
TLC plates, concentrating zone
These plates are based on different adsorption properties of two silica adsorbents: An inert large pore concentrating adsorbentMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable