Molybdenum, powder, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ200

1.12254.0100
1.12254.0100EA 222 EUR
1.12254.0100
Molybdenum, powder, Sigma-Aldrich®
Molybdenum
Formula: Mo
Boiling Pt: 5560 °C (1013 hPa)
Melting Pt: 2622 °C
Density: 10,2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003465
CAS Number: 7439-98-7
EINECS: 231-107-2
UN: 3089
ADR: 4.1,III

Order Now

Specification Test Results

Hill Formula Mo
Molar Mass 95.94 g/mol
Density 10.2 g/cm³ (20 °C)
Melting Point 2620 °C
Bulk density 1400 kg/m³
Solubility (20 °C) insoluble
Storage class 10 - 13 Other liquids and solids
WGK NWG not water endangering


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR