HPTLC and GLP plates, silica gel 60, unmodified layers, glass backed
Supplier: Merck
1.05564.0001 1.05616.0001 1.05628.0001 1.05629.0001 1.05631.0001 1.05633.0001 1.05641.0001 1.05642.0001 1.11764.0001 1.12363.0001 1.13187.0001 1.13727.0001 1.13728.0001 1.13748.0001 1.15498.0001 1.15552.0001 1.15696.0001 1.05648.0001 1.05649.0001 1.15534.0001
1.05564.0001EA 317 EUR
1.05564.0001 1.05616.0001 1.05628.0001 1.05629.0001 1.05631.0001 1.05633.0001 1.05641.0001 1.05642.0001 1.11764.0001 1.12363.0001 1.13187.0001 1.13727.0001 1.13728.0001 1.13748.0001 1.15498.0001 1.15552.0001 1.15696.0001 1.05648.0001 1.05649.0001 1.15534.0001
HPTLC and GLP plates, silica gel 60, unmodified layers, glass backed
TLC Plates
HPTLC plates, unmodified silica gel 60
This range of plates uses an optimised Merck silica gel 60 with a particle size of only 5 to 6 µm, which results in higher packing density and a smoother surface, therefore, reducing band diffusion. Less sample is required, 0,1 to 0,5 µl than for classic TLC.
  • Faster analysis, only 3 to 20 minutes for optimum separations
  • 5- to 10-fold increased sensitivity compared to classic TLC
  • Extra thin (100 µm) HPTLC automated multiple development (AMD) plates
HPTLC plates, concentrating zone
HPTLC concentrating-zone platesMore Product Information