Petroleum spirit, 40…60 °C, Suprasolv® hypergrade Trace_analysis_organic, Supelco®

Supplier: Merck
Danger

Synonyms: Gasoline, Petroleum ether, Petrol, Benzine

MQ100

1.16740.1000 1.16740.2500
1.16740.1000EA 88.9 EUR
1.16740.1000 1.16740.2500
Petroleum spirit, 40…60 °C, Suprasolv® hypergrade Trace_analysis_organic, Supelco®
Petroleum spirit

SupraSolv® ECD and FID is specially developed for gas chromatography in combination with ECD (Electron Capture Detetor) and FID (Flame Ionization detector). SupraSolv® solvents offer the largest specified retention time range, a clear baseline and a minimal signal-to-noise ratio leading to reliable and reproducible analysis results.

Boiling Pt: 40…60 °C (1013 hPa)
Density: 0.65 g/cm³ (20 °C)
Flash Pt: –40 °C
Storage Temperature: Ambient
MDL Number: MFCD00081849
CAS Number: 64742-49-0
EINECS: 265-151-9
UN: 1268
ADR: 3

Order Now

Specification Test Results

Boiling range (40-60°C) ≥ 90 % (v/v)
Evaporation residue ≤ 3.0 mg/l
Water ≤ 0.005 %
Color ≤ 10 Hazen
GC/ECD (retention range 1,2,4-tri- chlorobenzene to decachlorobiphenyle, individual signals (lindane standard)) ≤ 2 pg/ml
GC/FID (retention range n-undecane to n-tetracontane, individual signals (n-tetradecane standard)) ≤ 2 ng/ml
GC/MSD (retention range n-undecane to n-tetracontane, scaning area 30 - 600 amu, individual signals (n- tetradecane standard)) ≤ 2 ng/ml


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR