HPLC columns, Superspher®
Supplier: Merck
1.16009.0001 1.16010.0001 1.16051.0001 1.16052.0001 1.16054.0001 1.16056.0001 1.16854.0001 1.16855.0001 1.16857.0001 1.16858.0001 1.16869.0001 1.50193.0001 1.50198.0001 1.50791.0001 1.50792.0001 1.50973.0001 1.50974.0001 1.50975.0001 1.50980.0001 1.51299.0001 1.51909.0001 1.51910.0001 1.51299.1003 1.16010.1003 1.16051.1003 1.16052.1003 1.50973.1003
1.16009.0001EA 914 EUR
1.16009.0001 1.16010.0001 1.16051.0001 1.16052.0001 1.16054.0001 1.16056.0001 1.16854.0001 1.16855.0001 1.16857.0001 1.16858.0001 1.16869.0001 1.50193.0001 1.50198.0001 1.50791.0001 1.50792.0001 1.50973.0001 1.50974.0001 1.50975.0001 1.50980.0001 1.51299.0001 1.51909.0001 1.51910.0001 1.51299.1003 1.16010.1003 1.16051.1003 1.16052.1003 1.50973.1003
HPLC columns, Superspher®
Chromatography Columns
Superspher® improves the separation efficiency of HPLC analysis. This high performance spherical silica carrier, with a mean particle size of 4 μm, yields the best pressure/separation performance ratio in view of today's generation of HPLC systems and according to theoretical calculation and practical experience.
The number of theoretical plates for Superspher® is <100000 N/m. Therefore, they are always first choice if complex mixtures demand high peak capacity.
A broad range of modifications on Superspher® are available: Non polar derivatives (RP-8, RP-8 endcapped, RP-18, RP-18More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable