Visual colour kits, carbon dioxide, MQuant®
Supplier: Merck
1.17179.0001
1.17179.0001EA 148 EUR
1.17179.0001
Visual colour kits, carbon dioxide, MQuant®
Environmental Test Kits
Carbon dioxide is an important parameter for the assessment of the surface water quality and of the corrosive behavior of water. The kit is used for water and wastewater treatments following titrimetric method.
The consumption of titration solution is dependent on the concentration of the substance to be determined.
Delivery information: Kit includes a bottle of reagent CO2-1 (indicator solution); 2 bottles of reagent CO2-2 (titration solution), a measuring beaker, one graduated test vessel and a card with brief instruction.