Multicolour effect pigments, Colorstream®
Supplier: Merck
1.40648.1000 1.40648.9015
1.40648.1000EA 0 EUR
1.40648.1000 1.40648.9015
Multicolour effect pigments, Colorstream®
Coating Testers
Colorstream® multicolor effect pigments add dimensions to styling possibilities: They can change colour with the angle of observation, allowing innovative shades and new designs with that extra something.
  • Limitless styling possibilities with ever-changing highlights
  • Iridescent colour gradients
  • Unexpected changes that capture attention
Suitable for flexo/offset coating, gravure, screen and for all plastic applications. Available with a special weather treatment, they add that special something to cars and architecture and more.