HPLC column accessories
Supplier: Merck
1.50082.0001 1.50083.0001 1.50226.0001 1.50227.0001 1.51257.0001 1.51258.0001 1.51419.0001 1.50017.0001 1.51487.0001 1.51486.0001 1.51496.0001
1.50082.0001EA 441 EUR
1.50082.0001 1.50083.0001 1.50226.0001 1.50227.0001 1.51257.0001 1.51258.0001 1.51419.0001 1.50017.0001 1.51487.0001 1.51486.0001 1.51496.0001
HPLC column accessories
Chromatography Supplies Chromatography Spare Parts
Merck columns come in three formats.
1. The Hibar® RT is a traditional column with standard ¹/₁₆" female end fittings. The Hibar® stand-alone guard cartridge holder, in conjunction with the LiChrospher® 4-4 guard cartridges, can be used to protect the column.
2. The LiChroCART® cartridge columns utilise the manuCART® removable end fittings, which have standard ¹/₁₆" female connections. ManuCART® end fittings incorporate a guard cartridge holder as an integral part of the assembly which minimises dead volume. It can be used with any Merck column between 75 and 250 mm in length.
Order Now
SPECIFICATIONS
Environmentally Preferable