HPLC columns, LiChroCART®, Superspher® 60 RP-select B
Supplier: Merck
1.50173.7141 1.50178.7141 1.50179.7141 1.50195.7141 1.50228.7141 1.50229.7141 1.50233.7141 1.50234.7141 1.50236.7141 1.50302.7141 1.51431.7141 1.51432.7141 1.51442.7141 1.51443.7141 1.51444.7141 1.51448.7141 1.50170.7141 1.50201.7141 1.50175.7141
1.50173.7141EA 541 EUR
1.50173.7141 1.50178.7141 1.50179.7141 1.50195.7141 1.50228.7141 1.50229.7141 1.50233.7141 1.50234.7141 1.50236.7141 1.50302.7141 1.51431.7141 1.51432.7141 1.51442.7141 1.51443.7141 1.51444.7141 1.51448.7141 1.50170.7141 1.50201.7141 1.50175.7141
HPLC columns, LiChroCART®, Superspher® 60 RP-select B
Chromatography Columns
Stainless steel LiChroCART® cartridges are available with various types of Superspher® sorbents.
These LiChroCART® cartridges require part number 1.51486.0001 manu-CART® cartridge column holder, which can be used to hold one cartridge column with or without a  4×4 mm guard column. LiChroCART® columns 250×10 mm require part number 1.51419.0001 manu-CART® 10. The short LiChroCART® columns can be ordered as a set including the corresponding cartridge holder and one cartridge, or as a pack of 3 cartridges without cartridge holder. The separate part numbers for the cartridge are asMore Product Information