HPLC columns, Purospher® STAR RP-18 endcapped
Supplier: Merck
1.50036.0001 1.50037.0001 1.50225.0001 1.50231.0001 1.50239.0001 1.50240.0001 1.50241.0001 1.50242.0001 1.50250.0001 1.50251.0001 1.50252.0001 1.50253.0001 1.50254.0001 1.50255.0001 1.50256.0001 1.50358.0001 1.50359.0001 1.51455.0001 1.51456.0001 1.51460.0001 1.51448.7184 1.50596.0001 1.50615.0001 1.50620.0001 1.50414.0001 1.50617.0001 1.50470.0001 1.50431.0001 1.50469.0001 1.50471.0001 1.50622.0001 1.50427.0001 1.50468.0001 1.50626.0001 1.50398.0001 1.50413.0001 1.50627.0001 1.50607.0001 1.50621.0001 1.50593.0001 1.50625.0001 1.50624.0001 1.50612.0001 1.50428.0001 1.51914.0001 1.50037.1003 1.50251.1003 1.50252.1003 1.50358.1003 1.50359.1003 1.50431.1003 1.50617.1003 1.51455.1003 1.50036.1003 1.51456.1003 1.50393.0001
1.50036.0001EA 753 EUR
1.50036.0001 1.50037.0001 1.50225.0001 1.50231.0001 1.50239.0001 1.50240.0001 1.50241.0001 1.50242.0001 1.50250.0001 1.50251.0001 1.50252.0001 1.50253.0001 1.50254.0001 1.50255.0001 1.50256.0001 1.50358.0001 1.50359.0001 1.51455.0001 1.51456.0001 1.51460.0001 1.51448.7184 1.50596.0001 1.50615.0001 1.50620.0001 1.50414.0001 1.50617.0001 1.50470.0001 1.50431.0001 1.50469.0001 1.50471.0001 1.50622.0001 1.50427.0001 1.50468.0001 1.50626.0001 1.50398.0001 1.50413.0001 1.50627.0001 1.50607.0001 1.50621.0001 1.50593.0001 1.50625.0001 1.50624.0001 1.50612.0001 1.50428.0001 1.51914.0001 1.50037.1003 1.50251.1003 1.50252.1003 1.50358.1003 1.50359.1003 1.50431.1003 1.50617.1003 1.51455.1003 1.50036.1003 1.51456.1003 1.50393.0001
HPLC columns, Purospher® STAR RP-18 endcapped
Chromatography Columns
Fast method development of complex samples across the pH range from 1,5 to 10,5 with different mobile phases and temperature conditions.
Purospher® STAR RP-18 endcapped HPLC columns are designed for universal use. Basic, neutral and metal-chelating compounds can easily be separated with simple mobile phases – naturally without peak tailing. Thanks to its outstanding performance and stability, Purospher® STAR RP-18 endcapped offers maximum flexibility in method development. Robust methods can be developed over the entire pH range from 1,5 to 10,5. High pH-stability allows the separation ofMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable