HPLC columns, SeQuant® ZIC®-cHILIC
Supplier: Merck
1.50662.0001 1.50670.0001 1.50658.0001 1.50659.0001 1.50656.0001 1.50657.0001 1.50660.0001 1.50661.0001 1.50669.0001 1.50658.1003
1.50662.0001EA 1670 EUR
1.50662.0001 1.50670.0001 1.50658.0001 1.50659.0001 1.50656.0001 1.50657.0001 1.50660.0001 1.50661.0001 1.50669.0001 1.50658.1003
HPLC columns, SeQuant® ZIC®-cHILIC
Chromatography Columns
ZIC®-cHILIC is based on the same technology platform as ZIC®-HILIC, but has a different functional group and an improved manufacturing procedure. The different charge orientation of the zwitterionic functional group leads to a different – and complementary – selectivity for polar hydrophilic compounds. This selectivity is especially beneficial for hydrophilic compounds that are negatively charged, positively charged, zwitterionic or neutral, for example, organic acids, amino acids, metabolites, aminoglycosides and nucleotides.
  • Complementary selectivity by reoriented charges in
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable