HPLC columns, LiChrosorb®
Supplier: Merck
1.50318.0001 1.50332.0001 1.50333.0001 1.50334.0001 1.50388.0001 1.50432.0001 1.50433.0001 1.51343.0001 1.51345.0001 1.51349.0001 1.51351.0001 1.51353.0001 1.51354.0001 1.51355.0001 1.51356.0001 1.51902.0001 1.51903.0001
1.50318.0001EA 830 EUR
1.50318.0001 1.50332.0001 1.50333.0001 1.50334.0001 1.50388.0001 1.50432.0001 1.50433.0001 1.51343.0001 1.51345.0001 1.51349.0001 1.51351.0001 1.51353.0001 1.51354.0001 1.51355.0001 1.51356.0001 1.51902.0001 1.51903.0001
HPLC columns, LiChrosorb®
Chromatography Columns
LiChrosorb® is one of the most successful HPLC packing materials. It is a totally porous, irregular particle silica gel produced with pore sizes of 60 and 100 Å. LiChrosorb® HPLC columns are available either as a cartridge, requiring re-usable endfittings, or as a complete end-fitted column.
If guard columns are required, use LiChrospher® guard cartridges listed in LiChrospher® section.