Purospher® STAR RP-8 HPLC Columns, Endcapped
Supplier: Merck
1.50031.0001 1.50032.0001 1.50033.0001 1.50035.0001 1.50038.0001 1.50270.0001 1.50271.0001 1.50272.0001 1.50274.0001 1.51453.0001 1.51454.0001 1.50171.7220 1.51916.0001 1.51917.0001
1.50031.0001EA 745 EUR
1.50031.0001 1.50032.0001 1.50033.0001 1.50035.0001 1.50038.0001 1.50270.0001 1.50271.0001 1.50272.0001 1.50274.0001 1.51453.0001 1.51454.0001 1.50171.7220 1.51916.0001 1.51917.0001
Purospher® STAR RP-8 HPLC Columns, Endcapped
Chromatography Columns
Purospher® STAR RP-8 endcapped, like Purospher® STAR RP-18 endcapped, is based on high purity silica and has an almost complete surface coverage. Thus, Purospher® STAR RP-8 provides excellent peak symmetry for acidic, basic and even chelating compounds, highest column efficiency in terms of the number of theoretical plates, and exceptional stability from pH 1,5 to 10,5. In addition, they are ideal for a wide range of applications. As the sorbent is less hydrophobic than Purospher® STAR RP-18 endcapped, analytes will typically elute faster on the C8 phase. Purospher® STAR RP-8 endcappedMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable