Potassium permanganate solution 0,002 mol/l (0,01 N) (20°C), Titripur®, Supelco®

Supplier: Merck

Synonyms: Permanganic acid potassium salt

1.60322.1003
1.60322.1003EA 64.6 EUR
1.60322.1003
Potassium permanganate solution 0,002 mol/l (0,01 N) (20°C), Titripur®, Supelco®
Potassium permanganate

Merck have introduced into the range of Titripur® Titrants some packs with additional technology - Supelco® SmartTitrants that use an RFID tag to provide a seamless data transfer between the smart titrator and the RFID tag on the label of the bottle of titrant. It will ensure that all information needed for method execution, result calculation and documentation is transferred to the titrator equipped with this technology. MettlerToledo Compact and Excellence titrators include this technology. Please see ordering table below for packs with RFID tag.

Formula: KMnO₄
MW: 158.03 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011364
CAS Number: 7722-64-7
Merck Index: 12,07824

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR