Tin standard solution, 1000 mg/L Sn in 2 M hydrochloric acid (from SnCl₄) Certipur® standard for AAS, Supelco®
Supplier: Merck
1.70242.0100 1.70242.0500
1.70242.0100EA 52.3 EUR
1.70242.0100 1.70242.0500
Tin standard solution, 1000 mg/L Sn in 2 M hydrochloric acid (from SnCl₄) Certipur® standard for AAS, Supelco®
Standards Cationic Standards Tin Standards
Certifications: Traceable to SRM from NIST.