Buffer solutions, according to EP and USP, Certipur®
Supplier: Merck
1.09406.0500 1.99011.0001 1.99015.0500 1.99017.0001 1.99068.0500 1.99091.0500
1.09406.0500EA 54.1 EUR
1.09406.0500 1.99011.0001 1.99015.0500 1.99017.0001 1.99068.0500 1.99091.0500
Buffer solutions, according to EP and USP, Certipur®
Buffers pH Calibration Buffers
Buffer solutions intended for extremely precise calibration, qualification and monitoring of pH instruments, and for testing other buffer solutions.
  • Measured using 5-point calibration against NIST & PTB primary certified reference materials
  • Detailed certificate of analysis for every product
  • Solutions in packs of single use sachets (30×30 ml) help prevent contamination