Ice bucket, with lid
Supplier: KARTELL
1000
1000.EA 337 EUR
1000.
Ice bucket, with lid
Ice Containers
Made of red PUR. Equipped with lid and two side-handles. Can be used with dry or minced ice and cooling mixtures.
  • Thickness of about 15 mm
  • Excellent isolating feature
  • Liquid nitrogen resistant for a few minutes