Molybdenum ≥99.95% (metals basis), powder APS 3-7 µm

Supplier: Thermo Fisher Scientific
Warning

10030.18 10030.30 10030.A1 10030.06
10030.18EA 63 EUR
10030.18 10030.30 10030.A1 10030.06
Molybdenum ≥99.95% (metals basis), powder APS 3-7 µm
Molybdenum

Purity: 99.95% CAS No.: 7439-98-7 Appearance: Grey Odour: Odourless UN No.: 3089 MDL No.: MFCD00003465

Formula: Mo
Boiling Pt: 5560 °C (1013 hPa)
Melting Pt: 2622 °C
Density: 10,2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003465
CAS Number: 7439-98-7
EINECS: 231-107-2
UN: 3089
ADR: 4.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR