Niobium ≥99.8% (metals basis), powder -325 mesh

Supplier: Thermo Fisher Scientific
Danger

10275.14 10275.22 10275.36 10275.88
10275.14EA 98.6 EUR
10275.14 10275.22 10275.36 10275.88
Niobium ≥99.8% (metals basis), powder -325 mesh
Niobium

Purity: 99.8% CAS No.: 7440-03-1 Odour: Odourless UN No.: 3089 MDL No.: MFCD00011126

Formula: Nb
Storage Temperature: Ambient
MDL Number: MFCD00011126
CAS Number: 7440-03-1
EINECS: 231-113-5
UN: 3089
ADR: 4.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR