Thulium ≥99.9% (REO, rare earth oxide basis), foil, Thickness: 0.1 mm (0.004 in)

Supplier: Thermo Fisher Scientific
Danger


Packaged under argon.

10374.FIEA 1300 EUR
10374.FI
Thulium ≥99.9% (REO, rare earth oxide basis), foil, Thickness: 0.1 mm (0.004 in)
Thulium

Purity: 99.9% CAS No.: 7440-30-4 Physical Form: ≈0.58g/25x25mm, Packaged under argon Appearance: Silver-grey Odour: Odourless UN No.: 3178 MDL No.: MFCD00011281

Formula: Tm
MW: 168.93 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011281
CAS Number: 7440-30-4
EINECS: 231-140-2
UN: 3178
ADR: 4.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR