Mixed cellulose ester membranes, sterile, 0,2 µm, Whatman™
Supplier: Whatman products (Cytiva)
7187-114 10401770 10401772 10406970 10406972 10408714 10408712
7187114.EA 144 EUR
7187114. 10401770. 10401772. 10406970. 10406972. 514-1156 511-0138
Mixed cellulose ester membranes, sterile, 0,2 µm, Whatman™
Filters Membrane Filters
Whatman sterile MCE membrane filters from Cytiva's business are plain or gridded filters designed to support routine cell counting and particle detection.
  • Excellent contrast for easy particle detection
  • High internal surface area for sample adsorption
  • High dirt loading capacity
  • Biologically inert with good thermal stability
  • Uniform microporous structure delivers high flow rates
These MCE membrane filters are composed of cellulose acetate and cellulose nitrate, giving the membrane a more uniform surface than pure nitrocellulose filters. Other sterileMore Product Information