Cobalt ≥99.5% (metals basis), powder 1.6 µm

Supplier: Thermo Fisher Scientific
Danger

10455.09 10455.22 10455.36
10455.09EA 98.1 EUR
10455.09 10455.22 10455.36
Cobalt ≥99.5% (metals basis), powder 1.6 µm
Cobalt

Purity: 99.8% CAS No.: 7440-48-4 Appearance: Black Odour: Odourless UN No.: 3089 MDL No.: MFCD00010935

Formula: Co
MW: 58.93 g/mol
Boiling Pt: 2870
Melting Pt: 1495
Storage Temperature: Ambient
MDL Number: MFCD00010935
CAS Number: 7440-48-4
EINECS: 231-158-0
UN: 3089
ADR: 4.1 II
Merck Index: 12,02488

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR