Palladium(II) acetate (45,9 - 48,4% Pd)

Supplier: Thermo Fisher Scientific
Danger

10516.03 10516.06 10516.14
10516.03EA 190 EUR
10516.03 10516.06 10516.14
Palladium(II) acetate (45,9 - 48,4% Pd)
Palladium (II) acetate

CAS No.: 3375-31-3 Molecular Formula: C12H18O12Pd3 Formula Weight: 673.46 Physical Form: Needles Appearance: Red-brown Melting Point: ca 205° dec. MDL No.: MFCD00012453


Carbonylation, oxidation and C-C bond formation

Formula: C₁₂H₁₈O₁₂Pd₃
Melting Pt: ∼ 205 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012453
CAS Number: 3375-31-3
EINECS: 222-164-4
Merck Index: 14,6991

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR